missing translation for 'onlineSavingsMsg'
Learn More

Fluconazole N-Oxide, TRC

Product Code. 30134897
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134897 10 mg 10mg
17979829 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30134897 Supplier Toronto Research Chemicals Supplier No. TRCF42100510MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1997296-62-4
Molecular Formula C13H12F2N6O2
Molecular Weight (g/mol) 322.27
Synonym Fluconazole mono-N-Oxide
SMILES OC(Cn1cncn1)(Cn2c[n+]([O-])cn2)c3ccc(F)cc3F

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C13H12F2N6O2/c14-10-1-2-11(12(15)3-10)13(22,4-19-7-16-6-17-19)5-20-9-21(23)8-18-20/h1-3,6-9,22H,4-5H2
Formula Weight 322.099
Chemical Name or Material Fluconazole N-Oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.