CAS RN 19983-44-9
IUPAC Name:
6-nitro-1λ⁶-benzothiophene-1,1-dione
Synonyms:
6-nitro-1λ⁶-benzothiophene-1,1-dione
Molecular Weight (g/mol):
211.19
Molecular Formula:
C8H5NO4S
InChi Key:
ZRRGOUHITGRLBA-UHFFFAOYSA-N
SMILES:
[O-][N+](=O)C1=CC2=C(C=CS2(=O)=O)C=C1