missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Stattic
GREENER_CHOICE

Product Code. 18064192
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18064192 10 mg 10mg
18023812 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18064192 Supplier Tocris Bioscience™ Supplier No. 2798/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective STAT3 inhibitor

Small molecule inhibitor of STAT3. Inhibits binding of tyrosine-phosphorylated peptide motifs to STAT3 SH2 domain and inhibits STAT3 activation, dimerization and nuclear translocation. Displays selectivity over STAT1, STAT5, c-Myc/Max, Jun/Jun and Lck. Induces apoptosis in STAT3-dependent cancer cell lines.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19983-44-9
Molecular Formula C8H5NO4S

Specifications

Quantity 10 mg
Formula Weight 211.19
Purity >99%
Molecular Weight (g/mol) 211.191
Chemical Name or Material Stattic
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.