CAS RN 20012-10-6
CAS RN 20012-10-6
IUPAC Name:
1-phenyl-2,3,4,5-tetrahydro-1H-3-benzazepine-7,8-diol hydrobromide
Synonyms:
1-phenyl-2,3,4,5-tetrahydro-1H-3-benzazepine-7,8-diol hydrobromide
Molecular Weight (g/mol):
336.23
Molecular Formula:
C16H18BrNO2
InChi Key:
INNWVRBZMBCEJI-UHFFFAOYNA-N
SMILES:
Br.OC1=C(O)C=C2C(CNCCC2=C1)C1=CC=CC=C1