CAS RN 2006277-78-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-Succinimidyl 3-Methoxy-4-nitrobenzoate, TRC
CAS: 2006277-78-5 Molecular Formula: C12H10N2O7 Molecular Weight (g/mol): 294.22 IUPAC Name: (2,5-dioxopyrrolidin-1-yl) 3-methoxy-4-nitrobenzoate SMILES: COc1cc(ccc1[N+](=O)[O-])C(=O)ON2C(=O)CCC2=O