CAS RN 202865-72-3
IUPAC Name:
1-bromo-4-fluoro-2-iodobenzene
Synonyms:
1-bromo-4-fluoro-2-iodobenzene
Molecular Weight (g/mol):
300.9
Molecular Formula:
C6H3BrFI
InChi Key:
QDCBSSUCEUMLGC-UHFFFAOYSA-N
SMILES:
FC1=CC(I)=C(Br)C=C1
1
–
1
of
1
results
1
1-Bromo-4-fluoro-2-iodobenzene, 98+%
CAS: 202865-72-3 Molecular Formula: C6H3BrFI Molecular Weight (g/mol): 300.897 MDL Number: MFCD00042578 InChI Key: QDCBSSUCEUMLGC-UHFFFAOYSA-N Synonym: 2-bromo-5-fluoroiodobenzene,4-fluoro-2-iodobromobenzene,2-iodo-4-fluorobromobenzene,1-bromo-4-fluoro-2-iodo-benzene,benzene, 1-bromo-4-fluoro-2-iodo,2-bromo-5-fluoro-1-iodobenzene,pubchem3233,acmc-1cljm,ksc493k3j,1-bromo-2-iodo-4-fluorobenzene PubChem CID: 2773370 IUPAC Name: 1-bromo-4-fluoro-2-iodobenzene SMILES: C1=CC(=C(C=C1F)I)Br