Learn More
1-Bromo-4-fluoro-2-iodobenzene, 98+%
Product Code. 11490868
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11490868 25 g 25g
11470868 1 g 1g
11480868 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11490868 Supplier Thermo Scientific Alfa Aesar Supplier No. B25289.14
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
CAS: 202865-72-3 | C6H3BrFI | 300.897 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 202865-72-3
Molecular Formula C6H3BrFI
Molecular Weight (g/mol) 300.897
MDL Number MFCD00042578
InChI Key QDCBSSUCEUMLGC-UHFFFAOYSA-N
Synonym 2-bromo-5-fluoroiodobenzene, 4-fluoro-2-iodobromobenzene, 2-iodo-4-fluorobromobenzene, 1-bromo-4-fluoro-2-iodo-benzene, benzene, 1-bromo-4-fluoro-2-iodo, 2-bromo-5-fluoro-1-iodobenzene, pubchem3233, acmc-1cljm, ksc493k3j, 1-bromo-2-iodo-4-fluorobenzene
PubChem CID 2773370
IUPAC Name 1-bromo-4-fluoro-2-iodobenzene
SMILES C1=CC(=C(C=C1F)I)Br
Specifications
Density 2.3
Boiling Point 235°C
Flash Point >110°C (230°F)
Refractive Index 1.63
Quantity 25 g
Sensitivity Light sensitive
Formula Weight 300.9
Percent Purity ≥98%
Chemical Name or Material 1-Bromo-4-fluoro-2-iodobenzene