accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

2H2H2H2H2H2H2H2H

CAS RN 2037-26-5

IUPAC Name: 1-(²H₃)methyl(²H₅)benzene
Synonyms: 1-(²H₃)methyl(²H₅)benzene
Molecular Weight (g/mol): 100.19
Molecular Formula: C7H8
InChi Key: YXFVVABEGXRONW-JGUCLWPXSA-N
SMILES: [2H]C1=C([2H])C([2H])=C(C([2H])=C1[2H])C([2H])([2H])[2H]
Quantity 
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
Grade 
  • (11)
Solvent or Matrix 
  • (1)
  • (1)
  • (1)
Concentration 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (11)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)