accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 204905-77-1

OSOO

CAS RN 204905-77-1

IUPAC Name: 2H,3H-thieno[3,4-b][1,4]dioxine-5-carbaldehyde
Synonyms: 2H,3H-thieno[3,4-b][1,4]dioxine-5-carbaldehyde
Molecular Weight (g/mol): 170.18
Molecular Formula: C7H6O3S
InChi Key: GNVXYRDVJKJZTO-UHFFFAOYSA-N
SMILES: O=CC1=C2OCCOC2=CS1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results