accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

S

CAS RN 205-43-6

IUPAC Name: 11-thiatetracyclo[8.7.0.0²,⁷.0¹²,¹⁷]heptadeca-1(10),2,4,6,8,12(17),13,15-octaene
Synonyms: 11-thiatetracyclo[8.7.0.0²,⁷.0¹²,¹⁷]heptadeca-1(10),2,4,6,8,12(17),13,15-octaene
Molecular Weight (g/mol): 234.32
Molecular Formula: C16H10S
InChi Key: XZUMOEVHCZXMTR-UHFFFAOYSA-N
SMILES: S1C2=C(C=CC=C2)C2=C1C=CC1=CC=CC=C21
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)