missing translation for 'onlineSavingsMsg'
Learn More

Benzo[b]naphtho[1,2-d]thiophene, 97%, BLD Pharm™

Product Code. 31073701
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
5 g
10 g
25 g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31073701 250 mg
31058886 5 g
31060223 25 g
31058902 100 mg
31058921 1 g
31065067 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 31073701 Supplier BLD Pharm Supplier No. BD115657250mg
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 205-43-6
Molecular Formula C16H10S
MDL Number MFCD00215942
Quantity 250 mg
SMILES C12=CC=CC=C1C3=C4C=CC=CC4=CC=C3S2
Molecular Weight (g/mol) 234.32
Percent Purity 97%
Chemical Name or Material Benzo[b]naphtho[1,2-d]thiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.