accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOOO

CAS RN 2051-95-8

IUPAC Name: 4-oxo-4-phenylbutanoic acid
Synonyms: 4-oxo-4-phenylbutanoic acid
Molecular Weight (g/mol): 178.19
Molecular Formula: C10H10O3
InChi Key: KMQLIDDEQAJAGJ-UHFFFAOYSA-N
SMILES: OC(=O)CCC(=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (5)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (2)
  • (5)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)