accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 206551-43-1

H3COSBOHHO

CAS RN 206551-43-1

IUPAC Name: (5-acetylthiophen-2-yl)boronic acid
Synonyms: 5-acetylthiophen-2-ylboronic acid
Molecular Weight (g/mol): 169.99
Molecular Formula: C6H7BO3S
InChi Key: DCNMATSPQKWETQ-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(S1)B(O)O
Molecular Weight (g/mol) 
  • (4)
  • (2)
Quantity 
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (1)
  • (1)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)