accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 20780-74-9

BrOOHN

CAS RN 20780-74-9

IUPAC Name: 7-bromo-2,3-dihydro-1H-indole-2,3-dione
Synonyms: 7-bromo-1H-indole-2,3-dione
Molecular Weight (g/mol): 226.03
Molecular Formula: C8H4BrNO2
InChi Key: OCVKSIWBTJCXPV-UHFFFAOYSA-N
SMILES: BrC1=CC=CC2=C1NC(=O)C2=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results