accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 209863-09-2

CH3OO

CAS RN 209863-09-2

IUPAC Name: 3'-methoxy-[1,1'-biphenyl]-4-carbaldehyde
Synonyms: 3'-methoxy-[1,1'-biphenyl]-4-carbaldehyde
Molecular Weight (g/mol): 212.25
Molecular Formula: C14H12O2
InChi Key: LHVLDSOAJZLBMM-UHFFFAOYSA-N
SMILES: COC1=CC=CC(=C1)C1=CC=C(C=O)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results