CAS RN 209991-64-0
CAS RN 209991-64-0
IUPAC Name:
2-[2-fluoro-4-(trifluoromethyl)phenyl]acetic acid
Synonyms:
[2-fluoro-4-(trifluoromethyl)phenyl]acetic acid
Molecular Weight (g/mol):
222.14
Molecular Formula:
C9H6F4O2
InChi Key:
LUIOBGWYVHPKRC-UHFFFAOYSA-N
SMILES:
OC(=O)CC1=C(F)C=C(C=C1)C(F)(F)F
1
–
1
of
1
results
1
2-Fluoro-4-(trifluoromethyl)phenylacetic acid, 97%, Thermo Scientific™
CAS: 209991-64-0 Molecular Formula: C9H6F4O2 Molecular Weight (g/mol): 222.14 MDL Number: MFCD00061186 InChI Key: LUIOBGWYVHPKRC-UHFFFAOYSA-N Synonym: 2-fluoro-4-trifluoromethyl phenylacetic acid,2-fluoro-4-trifluoromethyphenylacetic acid,2-2-fluoro-4-trifluoromethyl phenyl acetic acid,2-fluoro-4-trifluoromethyl phenyl acetic acid,benzeneacetic acid, 2-fluoro-4-trifluoromethyl,pubchem2710,pubchem8433,2-fluoro-4-trifluoromethyphenylaceticacid,acmc-1cisp PubChem CID: 3863567 IUPAC Name: 2-[2-fluoro-4-(trifluoromethyl)phenyl]acetic acid SMILES: OC(=O)CC1=C(F)C=C(C=C1)C(F)(F)F