missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-(trifluoromethyl)phenylacetic acid, 97%, Thermo Scientific™

Product Code. 11466020
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11466020 5 g 5g
11456020 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11466020 Supplier Thermo Scientific Alfa Aesar Supplier No. H26573.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Used in the field of nanoparticles. Used in laboratory as reagent.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 209991-64-0
Molecular Formula C9H6F4O2
Molecular Weight (g/mol) 222.14
MDL Number MFCD00061186
InChI Key LUIOBGWYVHPKRC-UHFFFAOYSA-N
Synonym 2-fluoro-4-trifluoromethyl phenylacetic acid, 2-fluoro-4-trifluoromethyphenylacetic acid, 2-2-fluoro-4-trifluoromethyl phenyl acetic acid, 2-fluoro-4-trifluoromethyl phenyl acetic acid, benzeneacetic acid, 2-fluoro-4-trifluoromethyl, pubchem2710, pubchem8433, 2-fluoro-4-trifluoromethyphenylaceticacid, acmc-1cisp
PubChem CID 3863567
IUPAC Name 2-[2-fluoro-4-(trifluoromethyl)phenyl]acetic acid
SMILES OC(=O)CC1=C(F)C=C(C=C1)C(F)(F)F

Specifications

Melting Point 78°C to 81°C
Quantity 5 g
Formula Weight 222.14
Percent Purity 97%
Chemical Name or Material 2-Fluoro-4-(trifluoromethyl)phenylacetic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.