accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOOFFF

CAS RN 212755-76-5

IUPAC Name: methyl 3-oxo-3-[4-(trifluoromethyl)phenyl]propanoate
Synonyms: methyl 3-oxo-3-[4-(trifluoromethyl)phenyl]propanoate
Molecular Weight (g/mol): 246.19
Molecular Formula: C11H9F3O3
InChi Key: NMJOILIYLOHXGP-UHFFFAOYSA-N
SMILES: COC(=O)CC(=O)C1=CC=C(C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results