accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CONOCH3

CAS RN 213193-32-9

IUPAC Name: 1-(6-methoxypyridin-3-yl)ethan-1-one
Synonyms: 1-(6-methoxypyridin-3-yl)ethanone
Molecular Weight (g/mol): 151.17
Molecular Formula: C8H9NO2
InChi Key: RYOQZXOVBJIUSX-UHFFFAOYSA-N
SMILES: COC1=NC=C(C=C1)C(C)=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results