accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COI

CAS RN 2142-70-3

IUPAC Name: 1-(2-iodophenyl)ethan-1-one
Synonyms: o-iodobenzoyl
Molecular Weight (g/mol): 246.05
Molecular Formula: C8H7IO
InChi Key: XDXCBCXNCQGZPG-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=CC=C1I
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)