accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHOHOHO

CAS RN 2144-08-3

IUPAC Name: 2,3,4-trihydroxybenzaldehyde
Synonyms: 2,3,4-trihydroxybenzaldehyde
Molecular Weight (g/mol): 154.12
Molecular Formula: C7H6O4
InChi Key: CRPNQSVBEWWHIJ-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=O)C(O)=C1O
Molecular Weight (g/mol) 
  • (1)
  • (7)
Quantity 
  • (1)
  • (1)
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (1)
  • (7)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)