accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CHNHO

CAS RN 214746-69-7

IUPAC Name: 1-phenyl-1-[4-(piperidin-1-yl)phenyl]prop-2-yn-1-ol
Synonyms: 1-phenyl-1-[4-(piperidin-1-yl)phenyl]prop-2-yn-1-ol
Molecular Weight (g/mol): 291.39
Molecular Formula: C20H21NO
InChi Key: AIBIEAJBGHJQJF-UHFFFAOYNA-N
SMILES: OC(C#C)(C1=CC=CC=C1)C1=CC=C(C=C1)N1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results