missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-1-[4-(1-piperidinyl)phenyl]-2-propyn-1-ol, 97%

Product Code. 15470646
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15470646 25 g 25g
15460646 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15470646 Supplier Thermo Scientific Alfa Aesar Supplier No. H53435.14
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 214746-69-7 | C20H21NO | 291.394 g/mol

1-Phenyl-1-[4-(1-piperidinyl)phenyl]-2-propyn-1-ol is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Phenyl-1-[4-(1-piperidinyl)phenyl]-2-propyn-1-ol is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 214746-69-7
Molecular Formula C20H21NO
Molecular Weight (g/mol) 291.394
MDL Number MFCD17677347
InChI Key AIBIEAJBGHJQJF-UHFFFAOYSA-N
Synonym 1-phenyl-1-4-1-piperidinyl phenyl-2-propyn-1-ol, 1-phenyl-1-4-piperidin-1-yl phenyl prop-2-yn-1-ol, 1-phenyl-1-4-piperidinophenyl prop-2-yn-1-ol, 1-4-piperidinophenyl-1-phenyl-2-propyne-1-ol
PubChem CID 15536835
IUPAC Name 1-phenyl-1-(4-piperidin-1-ylphenyl)prop-2-yn-1-ol
SMILES C#CC(C1=CC=CC=C1)(C2=CC=C(C=C2)N3CCCCC3)O

Specifications

Melting Point 142°C to 144°C
Quantity 25 g
Formula Weight 291.39
Percent Purity 97%
Chemical Name or Material 1-Phenyl-1-[4-(1-piperidinyl)phenyl]-2-propyn-1-ol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.