CAS RN 2162-98-3
IUPAC Name:
1,10-dichlorodecane
Synonyms:
decane, 1,10-dichloro-
Molecular Weight (g/mol):
211.17
Molecular Formula:
C10H20Cl2
InChi Key:
RBBNTRDPSVZESY-UHFFFAOYSA-N
SMILES:
ClCCCCCCCCCCCl
1
–
2
of
2
results
2
1,10-Dichlorodecane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.