missing translation for 'onlineSavingsMsg'
Learn More

1,10-Dichlorodecane, 98%

Product Code. 10750751 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10750751 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10750751 Supplier Thermo Scientific Acros Supplier No. 255930250
 Restricted Item
Estimated Shipment: 23-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 2162-98-3 | C10H20Cl2 | 211.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 2162-98-3
CAS Min % 97.5
CAS Max % 100.0
Melting Point 15.6°C
Color Colorless
Density 0.9990g/mL
Boiling Point 275.0°C
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C10H20Cl2
Linear Formula Cl(CH2)10Cl
Refractive Index 1.4600 to 1.4620
MDL Number MFCD00000957
Quantity 25 g
Beilstein 01, 522
Specific Gravity 0.999
Synonym decane, 1,10-dichloro, decamethylene dichloride, decane,10-dichloro, 1, 10-dichlorodecane, acmc-1cbxv, ksc202q6p, 1,10-dichlorodecane, 1,10-decamethylene dichloride
Solubility Information Solubility in water: 0.77g/L (20 c)
InChI Key RBBNTRDPSVZESY-UHFFFAOYSA-N
SMILES ClCCCCCCCCCCCl
IUPAC Name 1,10-dichlorodecane
Molecular Weight (g/mol) 211.17
PubChem CID 75101
Viscosity 4.72 mPa.s (20°C)
Formula Weight 211.18
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1, 10-Dichlorodecane, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 1,10-Dichlorodecane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.