accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NBrClSOOCl

CAS RN 216394-05-7

IUPAC Name: 5-bromo-6-chloropyridine-3-sulfonyl chloride
Synonyms: 5-bromo-6-chloropyridine-3-sulfonyl chloride
Molecular Weight (g/mol): 290.94
Molecular Formula: C5H2BrCl2NO2S
InChi Key: TURGMVYIESHZBE-UHFFFAOYSA-N
SMILES: ClC1=C(Br)C=C(C=N1)S(Cl)(=O)=O
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)