missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-chloropyridine-5-sulfonyl chloride, 96%

Product Code. 11323906
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11323906 5 g 5g
11313906 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11323906 Supplier Thermo Scientific Alfa Aesar Supplier No. L17015.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 216394-05-7 | C5H2BrCl2NO2S | 290.94 g/mol

3-Bromo-2-chloropyridine-5-sulfonyl chloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromo-2-chloropyridine-5-sulfonyl chloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff field.

Solubility
Reacts with water. Soluble in dichloromethane.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under dry inert gas. It is sensitive to moisture. Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 216394-05-7
Molecular Formula C5H2BrCl2NO2S
Molecular Weight (g/mol) 290.94
MDL Number MFCD01318107
InChI Key TURGMVYIESHZBE-UHFFFAOYSA-N
Synonym 3-bromo-2-chloropyridine-5-sulfonyl chloride, 3-bromo-2-chloropyridine-5-sulphonyl chloride, 5-bromo-6-chloro-pyridine-3-sulfonyl chloride, 5-bromo-6-chloro-3-chlorosulphonyl pyridine, 5-bromo-6-chloro-3-pyridinesulfonyl chloride, 5-bromo-6-chloro-3-pyridine sulfonyl chloride, pubchem1289, 5-bromo-6-chloropyridine-3-sulfonylchloride, acmc-209xe4, 5-bromo-6-chloro 3-pyridyl chlorosulfone
PubChem CID 2793626
IUPAC Name 5-bromo-6-chloropyridine-3-sulfonyl chloride
SMILES ClC1=C(Br)C=C(C=N1)S(Cl)(=O)=O

Specifications

Melting Point 65°C to 70°C
Quantity 5 g
UN Number UN3261
Sensitivity Moisture sensitive
Solubility Information Reacts with water. Soluble in dichloromethane.
Formula Weight 290.95
Percent Purity 96%
Chemical Name or Material 3-Bromo-2-chloropyridine-5-sulfonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.