accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 216766-13-1

IUPAC Name: 5-methoxy-2-(trifluoromethyl)pyridine
Synonyms: 5-methoxy-2-(trifluoromethyl)pyridine
Molecular Weight (g/mol): 177.13
Molecular Formula: C7H6F3NO
InChi Key: MRHJZBSCOLVMGA-UHFFFAOYSA-N
SMILES: COC1=CN=C(C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results