accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 21679-31-2

IUPAC Name: tris(4-oxidaniumylidenepent-2-en-2-olate) chromium
Synonyms: chromium(III) acetylacetonate
Molecular Weight (g/mol): 352.35
Molecular Formula: C15H24CrO6
InChi Key: MJSNUBOCVAKFIJ-UHFFFAOYSA-N
SMILES: [Cr].CC([O-])=CC(C)=[OH+].CC([O-])=CC(C)=[OH+].CC([O-])=CC(C)=[OH+]
Physical Form 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results