accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClCl2H2H2H2H

CAS RN 2199-69-1

IUPAC Name: dichloro(²H₄)benzene
Synonyms: dichloro(²H₄)benzene
Molecular Weight (g/mol): 151.02
Molecular Formula: C6H4Cl2
InChi Key: RFFLAFLAYFXFSW-RHQRLBAQSA-N
SMILES: [2H]C1=C([2H])C([2H])=C(Cl)C(Cl)=C1[2H]
Molecular Weight (g/mol) 
  • (7)
  • (3)
Quantity 
  • (2)
  • (3)
  • (1)
  • (1)
  • (3)
Grade 
  • (3)
Percent Purity 
  • (1)
  • (2)
Solvent or Matrix 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
  • (7)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)