accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOOOOCH3

CAS RN 22027-52-7

IUPAC Name: methyl 4-(3-ethoxy-3-oxopropanoyl)benzoate
Synonyms: methyl 4-(3-ethoxy-3-oxopropanoyl)benzoate
Molecular Weight (g/mol): 250.25
Molecular Formula: C13H14O5
InChi Key: GFDCYWYWNSTMDX-UHFFFAOYSA-N
SMILES: CCOC(=O)CC(=O)C1=CC=C(C=C1)C(=O)OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results