accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOO

CAS RN 2210-74-4

IUPAC Name: 2-[(2-methoxyphenoxy)methyl]oxirane
Synonyms: 2-(2-methoxyphenoxymethyl)oxirane
Molecular Weight (g/mol): 180.2
Molecular Formula: C10H12O3
InChi Key: RJNVSQLNEALZLC-UHFFFAOYNA-N
SMILES: COC1=CC=CC=C1OCC1CO1
Molecular Weight (g/mol) 
  • (4)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results