CAS RN 22204-53-1
(S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid, 99%
CAS: 22204-53-1 Molecular Formula: C14H14O3 Molecular Weight (g/mol): 230.263 MDL Number: MFCD00010500 InChI Key: CMWTZPSULFXXJA-VIFPVBQESA-N Synonym: naproxen,s-naproxen,naprosyn,naproxene,+-naproxen,equiproxen,aleve,s-+-2-6-methoxy-2-naphthyl propionic acid,laraflex,naproxeno PubChem CID: 156391 ChEBI: CHEBI:7476 IUPAC Name: (2S)-2-(6-methoxynaphthalen-2-yl)propanoic acid SMILES: CC(C1=CC2=C(C=C1)C=C(C=C2)OC)C(=O)O
Naproxen, 99.3%, MP Biomedicals™
CAS: 22204-53-1 Molecular Formula: C14H14O3 Molecular Weight (g/mol): 230.263 InChI Key: CMWTZPSULFXXJA-VIFPVBQESA-N Synonym: naproxen,s-naproxen,naprosyn,naproxene,+-naproxen,equiproxen,aleve,s-+-2-6-methoxy-2-naphthyl propionic acid,laraflex,naproxeno PubChem CID: 156391 ChEBI: CHEBI:7476 IUPAC Name: (2S)-2-(6-methoxynaphthalen-2-yl)propanoic acid SMILES: CC(C1=CC2=C(C=C1)C=C(C=C2)OC)C(=O)O
Naproxen, MP Biomedicals
CAS: 22204-53-1 Molecular Formula: C14H14O3 Molecular Weight (g/mol): 230.263 InChI Key: CMWTZPSULFXXJA-VIFPVBQESA-N Synonym: naproxen,s-naproxen,naprosyn,naproxene,+-naproxen,equiproxen,aleve,s-+-2-6-methoxy-2-naphthyl propionic acid,laraflex,naproxeno PubChem CID: 156391 ChEBI: CHEBI:7476 IUPAC Name: (2S)-2-(6-methoxynaphthalen-2-yl)propanoic acid SMILES: CC(C1=CC2=C(C=C1)C=C(C=C2)OC)C(=O)O
Naproxen, Lipomed
Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.
Naproxen 1.0 mg/ml in Methanol, LoGiCal
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
Naproxen 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
Naproxen, LoGiCal
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
Naproxen, Lipomed
Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.
Naproxen, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(S)-Naproxen, TRC
CAS: 22204-53-1 Molecular Formula: C14 H14 O3 Molecular Weight (g/mol): 230.26 Synonym: 2-Naphthaleneacetic acid, 6-methoxy-α-methyl-, (αS)- (9CI),2-Naphthaleneacetic acid, 6-methoxy-α-methyl-, (+)- (8CI),2-Naphthaleneacetic acid, 6-methoxy-α-methyl-, (S)-,(+)-(S)-Naproxen,(+)-2-(6-Methoxy-2-naphthyl)propionic acid,(+)-6-Methoxy-α-methyl-2-naphthaleneacetic acid,(+)-Naproxen,(S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid,(S)-(+)-Naproxen,(S)-(+)-Naproxene,(S)-2-(6-Methoxy-2-naphthyl)propanoic acid,(S)-2-(6-Methoxy-2-naphthyl)propionic acid,(S)-6-Methoxy-α-methyl-2-naphthaleneacetic acid,(S)-Naproxen,Apo-Naproxen,Bonyl,CG 3117,Diocodal,Dysmenalgit,Equiproxen,Floginax,Laraflex,Laser,MNPA,Naixan,Napren,Naprium,Naprius,Naprosyn,Naprosyne,Naproxen,Naprux,Naxen,Nycopren,Panoxen,Prexan,Proxen,Proxine,RS 3540,Reuxen,Veradol,Xenar,d-2-(6-Methoxy-2-naphthyl)propionic acid,d-Naproxen IUPAC Name: (2S)-2-(6-methoxynaphthalen-2-yl)propanoic acid SMILES: COc1ccc2cc(ccc2c1)[C@H](C)C(=O)O
Naproxen, 1mg/ml in Methanol, Lipomed
Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.