CAS RN 2233-18-3
IUPAC Name:
4-hydroxy-3,5-dimethylbenzaldehyde
Synonyms:
4-hydroxy-3,5-dimethylbenzaldehyde
Molecular Weight (g/mol):
150.18
Molecular Formula:
C9H10O2
InChi Key:
UYGBSRJODQHNLQ-UHFFFAOYSA-N
SMILES:
CC1=CC(C=O)=CC(C)=C1O
1
–
1
of
1
results
1
4-Hydroxy-3,5-dimethylbenzaldehyde, 97+%
CAS: 2233-18-3 Molecular Formula: C9H10O2 Molecular Weight (g/mol): 150.18 MDL Number: MFCD00006946 InChI Key: UYGBSRJODQHNLQ-UHFFFAOYSA-N Synonym: 3,5-dimethyl-4-hydroxybenzaldehyde,benzaldehyde, 4-hydroxy-3,5-dimethyl,4-formyl-2,6-xylenol,4-hydroxy-3,5-dimethyl-benzaldehyde,unii-23pa2pmp9f,23pa2pmp9f,syringnldehyde,zlchem 510,pubchem8252,buttpark 9657-68 PubChem CID: 75222 IUPAC Name: 4-hydroxy-3,5-dimethylbenzaldehyde SMILES: CC1=CC(C=O)=CC(C)=C1O