accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3CH3I

CAS RN 22445-41-6

IUPAC Name: 1-iodo-3,5-dimethylbenzene
Synonyms: 1-iodo-3,5-dimethylbenzene
Molecular Weight (g/mol): 232.06
Molecular Formula: C8H9I
InChi Key: ZLMKEENUYIUKKC-UHFFFAOYSA-N
SMILES: CC1=CC(I)=CC(C)=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)