missing translation for 'onlineSavingsMsg'
Learn More

5-Iodo-m-xylene, 97%

Product Code. 11498337
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11498337 10 g 10g
11408347 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11498337 Supplier Thermo Scientific Alfa Aesar Supplier No. A18413.09
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 22445-41-6 | C8H9I | 232.06 g/mol

1-Iodo-3,5-dimethylbenzene (5-iodo-m-xylene) is suitable for use in the synthesis of N-(3,5-xylyl)-N-ethyl aniline, an aryl amine, It is also used in the following studies; α-Arylation of ketones, Copper-catalyzed N-arylation of imidazoles, Cyanation of 5-iodo-m-xylene to form 3,5-dimethylbenzonitrile, synthesis of 1,3-Dimethyl-5-phenoxybenzene by nano-CuFe2O4 catalyzed C-O cross-coupling with phenol, CuBr-catalyzed amination of 1-iodo-3,5-dimethylbenzene to form N-Allyl-3,5-dimethylbenzenamine, copper-catalyzed C-S bond-formation between 5-iodo-m-xylene and thiophenol. It is also employed as a starting material in the synthesis of biphenyl-3,3’,5,5’-tetracarboxylic acid, radical bromination of 5-iodo-m-xylene by N-bromosuccinimide to form 1,3-bis(bromomethyl)-5-iodobenzene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Iodo-3,5-dimethylbenzene (5-iodo-m-xylene) is suitable for use in the synthesis of N-(3,5-xylyl)-N-ethyl aniline, an aryl amine, It is also used in the following studies; α-Arylation of ketones, Copper-catalyzed N-arylation of imidazoles, Cyanation of 5-iodo-m-xylene to form 3,5-dimethylbenzonitrile, synthesis of 1,3-Dimethyl-5-phenoxybenzene by nano-CuFe2O4 catalyzed C-O cross-coupling with phenol, CuBr-catalyzed amination of 1-iodo-3,5-dimethylbenzene to form N-Allyl-3,5-dimethylbenzenamine, copper-catalyzed C-S bond-formation between 5-iodo-m-xylene and thiophenol. It is also employed as a starting material in the synthesis of biphenyl-3,3’,5,5’-tetracarboxylic acid, radical bromination of 5-iodo-m-xylene by N-bromosuccinimide to form 1,3-bis(bromomethyl)-5-iodobenzene.

Solubility
Sparingly soluble (0.014 g/L) (25°C), Calc.

Notes
Light Sensitive. Store away from oxidizing agents and light. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22445-41-6
Molecular Formula C8H9I
Molecular Weight (g/mol) 232.06
MDL Number MFCD00060659
InChI Key ZLMKEENUYIUKKC-UHFFFAOYSA-N
Synonym 5-iodo-m-xylene, 1,3-dimethyl-5-iodobenzene, benzene, 1-iodo-3,5-dimethyl, 3,5-dimethyliodobenzene, pubchem3872, acmc-1ccdf, 3,5-dimethyl-1-iodobenzene, ksc422q9l, 1-iodo-3,5-dimethylbenzene, #
PubChem CID 140924
IUPAC Name 1-iodo-3,5-dimethylbenzene
SMILES CC1=CC(I)=CC(C)=C1

Specifications

Density 1.61
Quantity 10 g
Refractive Index 1.595
Beilstein 2039353
Boiling Point 229°C
Solubility Information Sparingly soluble (0.014g/L) (25°C),Calc.
Formula Weight 232.07
Percent Purity 98%
Chemical Name or Material 5-Iodo-m-xylene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.