accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

SnH3CCH2CH3H2CCH3OCHCH3OCH3OCHCH3O

CAS RN 22673-19-4

IUPAC Name: tin(4+) bis(2,4-dioxopentan-3-ide) bis(pentan-1-ide)
Synonyms: tin(4+) bis(pentan-1-ide) diacetylacetonate
Molecular Weight (g/mol): 459.21
Molecular Formula: C20H36O4Sn
InChi Key: VJLXWWRKGIBBSL-UHFFFAOYSA-N
SMILES: [Sn+4].CCCC[CH2-].CCCC[CH2-].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results