accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H2OH2OHO(R)OH(R)OH(S)OH(R)OHOOHO(R)OH(R)OH(S)OH(R)OHOOFe

CAS RN 22830-45-1

IUPAC Name: λ²-iron(2+) bis((2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate) dihydrate
Synonyms: λ²-iron(2+) bis(D-gluconate) dihydrate
Molecular Weight (g/mol): 482.17
Molecular Formula: C12H26FeO16
InChi Key: OKGNXSFAYMSVNN-SYAJEJNSSA-L
SMILES: O.O.[Fe++].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results