missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) gluconate dihydrate, 94%

Product Code. 11480398
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11480398 500 g 500g
11470398 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11480398 Supplier Thermo Scientific Alfa Aesar Supplier No. B24561.36
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 22830-45-1 | C12H26FeO16 | 482.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22830-45-1
Molecular Formula C12H26FeO16
Molecular Weight (g/mol) 482.17
MDL Number MFCD00150872
InChI Key OKGNXSFAYMSVNN-SYAJEJNSSA-L
Synonym iron ii gluconate hydrate
PubChem CID 131851469
SMILES O.O.[Fe++].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O

Specifications

Melting Point ∼140°C
Quantity 500 g
IUPAC Name λ²-iron(2+) bis((2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate) dihydrate
Formula Weight 482.19 (446.19 Anhydrous)
Percent Purity 94%
Chemical Name or Material Iron(II) gluconate dihydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.