accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NO

CAS RN 22934-41-4

IUPAC Name: quinoline-5-carbaldehyde
Synonyms: quinoline-5-carbaldehyde
Molecular Weight (g/mol): 157.17
Molecular Formula: C10H7NO
InChi Key: BNEVFKZLYCGDFG-UHFFFAOYSA-N
SMILES: O=CC1=C2C=CC=NC2=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results