CAS RN 2304-30-5
IUPAC Name:
tetrabutylphosphanium chloride
Synonyms:
tetrabutylphosphonium chloride
Molecular Weight (g/mol):
294.89
Molecular Formula:
C16H36ClP
InChi Key:
IBWGNZVCJVLSHB-UHFFFAOYSA-M
SMILES:
[Cl-].CCCC[P+](CCCC)(CCCC)CCCC
1
–
1
of
1
results
1
Tetrabutylphosphonium Chloride, TRC
CAS: 2304-30-5 Molecular Formula: C16H36ClP Molecular Weight (g/mol): 294.88 Synonym: Cyphos 443P,Cyphos 443T,Cyphos IL 164,Hishicolin PX 4C,Tetra-n-butylphosphonium Chloride IUPAC Name: tetrabutylphosphanium;chloride SMILES: [Cl-].CCCC[P+](CCCC)(CCCC)CCCC