missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylphosphonium Chloride, TRC

Product Code. 30455867
Change view
Click to view available options
Quantity:
5 g
100 g
Unit Size:
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30455867 5 g 5g
30449521 100 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30455867 Supplier Toronto Research Chemicals Supplier No. TRCT3106705G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2304-30-5
Molecular Formula C16H36ClP
Molecular Weight (g/mol) 294.88
Synonym Cyphos 443P, Cyphos 443T, Cyphos IL 164, Hishicolin PX 4C, Tetra-n-butylphosphonium Chloride
IUPAC Name tetrabutylphosphanium;chloride
SMILES [Cl-].CCCC[P+](CCCC)(CCCC)CCCC

Specifications

Quantity 5 g
InChI Formula InChI=1S/C16H36P.ClH/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;/h5-16H2,1-4H3;1H/q+1;/p-1
Formula Weight 294.2243
Chemical Name or Material Tetrabutylphosphonium Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.