accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HClCH3H3CHNNS

CAS RN 23076-35-9

IUPAC Name: hydrogen N-(2,6-dimethylphenyl)-5,6-dihydro-4H-1,3-thiazin-2-amine chloride
Synonyms: hydrogen xylazine chloride
Molecular Weight (g/mol): 256.79
Molecular Formula: C12H17ClN2S
InChi Key: QYEFBJRXKKSABU-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CC1=CC=CC(C)=C1NC1=NCCCS1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)