missing translation for 'onlineSavingsMsg'
Learn More

Xylazine hydrochloride, Thermo Scientific™

Agonist at the alpha-2 adrenergic receptor

Brand:  Thermo Scientific Alfa Aesar J62394.06

Quantity 5g

 View more versions of this product

Product Code. 15464779

Request Bulk or Custom Format

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

23076-35-9
256.79
QYEFBJRXKKSABU-UHFFFAOYSA-N
68554
[H+].[Cl-].CC1=CC=CC(C)=C1NC1=NCCCS1
C12H17ClN2S
MFCD00058196
xylazine hydrochloride, xylazine hcl, n-2,6-dimethylphenyl-5,6-dihydro-4h-1,3-thiazin-2-amine hydrochloride, celactal, xylazine chloride, bay 1470 hydrochloride, unii-ngc3s0882s, ccris 8685, bay va 1470
hydrogen N-(2,6-dimethylphenyl)-5,6-dihydro-4H-1,3-thiazin-2-amine chloride

Specifications

23076-35-9
MFCD00058196
UN2811
14,10080
Soluble in water
[H+].[Cl-].CC1=CC=CC(C)=C1NC1=NCCCS1
256.79
256.79
C12H17ClN2S
5g
6448471
xylazine hydrochloride, xylazine hcl, n-2,6-dimethylphenyl-5,6-dihydro-4h-1,3-thiazin-2-amine hydrochloride, celactal, xylazine chloride, bay 1470 hydrochloride, unii-ngc3s0882s, ccris 8685, bay va 1470
QYEFBJRXKKSABU-UHFFFAOYSA-N
hydrogen N-(2,6-dimethylphenyl)-5,6-dihydro-4H-1,3-thiazin-2-amine chloride
68554
Xylazine hydrochloride
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.