missing translation for 'onlineSavingsMsg'
Learn More

Xylazine hydrochloride, Thermo Scientific™

Agonist at the alpha-2 adrenergic receptor

153.77€

Chemical Identifiers

CAS 23076-35-9
Molecular Formula C12H17ClN2S
Molecular Weight (g/mol) 256.79
MDL Number MFCD00058196
InChI Key QYEFBJRXKKSABU-UHFFFAOYSA-N
Synonym xylazine hydrochloride, xylazine hcl, n-2,6-dimethylphenyl-5,6-dihydro-4h-1,3-thiazin-2-amine hydrochloride, celactal, xylazine chloride, bay 1470 hydrochloride, unii-ngc3s0882s, ccris 8685, bay va 1470
PubChem CID 68554
IUPAC Name hydrogen N-(2,6-dimethylphenyl)-5,6-dihydro-4H-1,3-thiazin-2-amine chloride
SMILES [H+].[Cl-].CC1=CC=CC(C)=C1NC1=NCCCS1
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15464779
SDS View Documents
Thermo Scientific Alfa Aesar
J62394.06
5g This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

23076-35-9
256.79
QYEFBJRXKKSABU-UHFFFAOYSA-N
68554
[H+].[Cl-].CC1=CC=CC(C)=C1NC1=NCCCS1
C12H17ClN2S
MFCD00058196
xylazine hydrochloride, xylazine hcl, n-2,6-dimethylphenyl-5,6-dihydro-4h-1,3-thiazin-2-amine hydrochloride, celactal, xylazine chloride, bay 1470 hydrochloride, unii-ngc3s0882s, ccris 8685, bay va 1470
hydrogen N-(2,6-dimethylphenyl)-5,6-dihydro-4H-1,3-thiazin-2-amine chloride

Specifications

23076-35-9
MFCD00058196
6448471
xylazine hydrochloride, xylazine hcl, n-2,6-dimethylphenyl-5,6-dihydro-4h-1,3-thiazin-2-amine hydrochloride, celactal, xylazine chloride, bay 1470 hydrochloride, unii-ngc3s0882s, ccris 8685, bay va 1470
QYEFBJRXKKSABU-UHFFFAOYSA-N
hydrogen N-(2,6-dimethylphenyl)-5,6-dihydro-4H-1,3-thiazin-2-amine chloride
68554
Xylazine hydrochloride
C12H17ClN2S
UN2811
14,10080
Soluble in water
[H+].[Cl-].CC1=CC=CC(C)=C1NC1=NCCCS1
256.79
256.79
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H350-H315-H319-H335
Toxic if swallowed.
May cause cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

missing translation for 'rtecsNumber' : XJ0776350

Recommended Storage : −30° to −10°C

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.