accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H3CCH3SCH3H3CSOHCH3H3CCH3CH3CH3H3CHOH3CH3CCH3

CAS RN 23288-49-5

IUPAC Name: 2,6-di-tert-butyl-4-({2-[(3,5-di-tert-butyl-4-hydroxyphenyl)sulfanyl]propan-2-yl}sulfanyl)phenol
Synonyms: probucol lursell lurselle aventis brand of probucol probucol (substance) bisbid lorelco almirall brand of probucol hoechst brand of probucol superlipid
Molecular Weight (g/mol): 516.84
Molecular Formula: C31H48O2S2
InChi Key: FYPMFJGVHOHGLL-UHFFFAOYSA-N
SMILES: CC(C)(SC1=CC(=C(O)C(=C1)C(C)(C)C)C(C)(C)C)SC1=CC(=C(O)C(=C1)C(C)(C)C)C(C)(C)C
Molecular Weight (g/mol) 
  • (5)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (3)
  • (2)
Percent Purity 
  • (1)
  • (2)
Certifications/Compliance 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (2)
  • (3)
  • (5)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)