Learn More
4,4'-(Propane-2,2-diylbis(sulfanediyl))bis(2,6-di-tert-butylphenol), 98%, BLD Pharm™
Specifications
Specifications
| CAS | 23288-49-5 |
| Molecular Formula | C31H48O2S2 |
| MDL Number | MFCD00079281 |
| Quantity | 100 mg |
| SMILES | CC(SC1=CC(C(C)(C)C)=C(O)C(C(C)(C)C)=C1)(SC2=CC(C(C)(C)C)=C(O)C(C(C)(C)C)=C2)C |
| Molecular Weight (g/mol) | 516.84 |
| Percent Purity | 98% |
| Chemical Name or Material | 4,4'-(Propane-2,2-diylbis(sulfanediyl))bis(2,6-di-tert-butylphenol) |