accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 23537-25-9

NH3(R)SOOOOO

CAS RN 23537-25-9

IUPAC Name: (2R)-2-azaniumyl-3-sulfonatopropanoate
Synonyms: L-cysteate L-cysteate(1-) L-cysteate anion
Molecular Weight (g/mol): 168.14
Molecular Formula: C3H6NO5S
InChi Key: XVOYSCVBGLVSOL-REOHCLBHSA-M
SMILES: [NH3+][C@@H](CS([O-])(=O)=O)C([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)