missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ L-Cysteic acid monohydrate, 99%

Product Code. 11377246
Click to view available options
Quantity:
10g
Unit Size:
10g
This item is not returnable. View return policy

Product Code. 11377246

Brand: Alfa Aesar™ L09135.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy
Applications
L-Cysteic acid monohydrate is used as a competitive inhibitor of the bacterial aspartate:alanine antiporter (AspT) exchange of aspartate and in other aspartate biological systems. It is used in monomeric surfactant development. It is used in studies of excitatory amino acids in the brain, such as those that bind to cysteine sulfinic acid receptors. L-Cysteic acid is a useful agonist at several rat metabotropic glutamate receptors (mGluRs).

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Specifications

CAS 23537-25-9
Molecular Formula C3H6NO5S
MDL Number MFCD00149544
Quantity 10g
Beilstein 1725495
Merck Index 14,2780
Synonym l-cysteic acid monohydrate, unii-vn8x20t18r, cysteic acid monohydrate, l, r-2-amino-3-sulfopropanoic acid hydrate, l-alanine, 3-sulfo-, monohydrate, cysteinesulfonic acid hydrate, h-cys o 2-oh.h2o, l-cysteic acid hydrate, l-cysteic acid monohydrate mi, alanine, 3-sulfo-, monohydrate, l
InChI Key XVOYSCVBGLVSOL-REOHCLBHSA-M
Optical Rotation +8° (c=5 in Water)
SMILES [NH3+][C@@H](CS([O-])(=O)=O)C([O-])=O
IUPAC Name (2R)-2-azaniumyl-3-sulfonatopropanoate
Molecular Weight (g/mol) 168.14
PubChem CID 12308854
Formula Weight 187.19 (169.17 Anhydrous)
Percent Purity 99%
Chemical Name or Material L-Cysteic acid monohydrate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.