accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 23828-92-4

IUPAC Name: hydrogen 4-{[(2-amino-3,5-dibromophenyl)methyl]amino}cyclohexan-1-ol chloride
Synonyms: hydrogen ambroβ chloride
Molecular Weight (g/mol): 414.57
Molecular Formula: C13H19Br2ClN2O
InChi Key: QNVKOSLOVOTXKF-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NC1=C(Br)C=C(Br)C=C1CNC1CCC(O)CC1
Molecular Weight (g/mol) 
  • (2)
  • (13)
Percent Purity 
  • (3)
  • (7)
  • (2)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
Certifications/Compliance 
  • (2)
  • (1)
Industry Type 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (2)
  • (1)
  • (2)
  • (3)
  • (7)
  • (2)
  • (13)
  • (3)
  • (7)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)